Promotional price valid on web orders only. Your contract pricing may differ. Interested in signing up for a dedicated account number?
Learn More

Potassium Acetate, ≥99.0%, ACS Reagent, Solstice

Catalog No. 60045444 Shop All Solstice Research Chemicals Products
Change view
Click to view available options
Quantity:
100 g
500 g
2.5 kg
Packaging:
Poly Bottle
3 product options available for selection
Product selection table with 3 available options. Use arrow keys to navigate and Enter or Space to select.
Catalog No. Quantity Packaging
60-045-444 500 g Poly Bottle
60-045-442 100 g Poly Bottle
60-045-443 2.5 kg Poly Bottle
Use arrow keys to navigate between rows. Press Enter or Space to select a product option. 3 options available.
3 options
Catalog No. 60-045-444 Supplier Solstice Supplier No. 236497500G
Add to Cart
Add to Cart

missing translation for 'validMainframeMsg'

ACS Reagent,≥99.0%

  • Leading the Industry For Over 65 Years
  • Solstice now delivers premium grade inorganic reagents worldwide – with consistency, purity, and accuracy assured
TRUSTED_SUSTAINABILITY

Chemical Identifiers

CAS 127-08-2
Molecular Formula C2H3KO2
Molecular Weight (g/mol) 98.142
MDL Number MFCD00012458
InChI Key SCVFZCLFOSHCOH-UHFFFAOYSA-M
Synonym potassium acetate, acetic acid, potassium salt, diuretic salt, potassium ethanoate, koac, octan draselny czech, potassiumacetate, acetic acid potassium salt, acok, fema no. 2920
PubChem CID 517044
ChEBI CHEBI:32029
IUPAC Name potassium;acetate
SMILES CC(=O)[O-].[K+]

Specifications

Melting Point 292°C
Physical Form Solid
Quantity 500 g
Linear Formula CH3COOK
UN Number NONH for all modes of transport
Beilstein 3595449
Solubility Information Soluble in water, 98.2g/L at 20°C
Formula Weight 98.14g/mol
Percent Purity ≥99.0%
Grade ACS Reagent
Packaging Poly Bottle
Density 1.57g/cu.cm at 25°C
Chemical Name or Material Potassium Acetate
Show More Show Less
EINECSNumber 204-822-2
RTECSNumber AJ3325000
WARNING: Cancer and Reproductive Harm - www.P65Warnings.ca.gov
Product Title
Select an issue

By clicking Submit, you acknowledge that you may be contacted by Fisher Scientific in regards to the feedback you have provided in this form. We will not share your information for any other purposes. All contact information provided shall also be maintained in accordance with our Privacy Policy.