Promotional price valid on web orders only. Your contract pricing may differ. Interested in signing up for a dedicated account number?
Learn More

Potassium Acetate, ACS Reagent, Reagents

Catalog No. 01337877
Change view
Click to view available options
Quantity: 500 g
1 product options available for selection
Product selection table with 1 available options. Use arrow keys to navigate and Enter or Space to select.
Catalog No.
01-337-877 Quantity: 500 g
Use arrow keys to navigate between rows. Press Enter or Space to select a product option. 1 options available.
1 options
Catalog No. 01-337-877 Supplier Reagents Holdings Llc Supplier No. C1276000500A5
 Restricted Item
Add to Cart
Add to Cart

missing translation for 'validMainframeMsg'

Potassium Acetate, ACS Reagent, ACS, for general laboratory use. Manufactured in ISO 9001 facility.

Chemical Identifiers

CAS 127-08-2
Molecular Formula C2H3KO2
Molecular Weight (g/mol) 98.14
InChI Key SCVFZCLFOSHCOH-UHFFFAOYSA-M
Synonym Acetic Acid potassium salt
IUPAC Name potassium acetate
SMILES [K+].CC([O-])=O

Specifications

Melting Point 303°C to 307°C
Color Colorless/White
Physical Form Solid
Quantity 500 g
Grade ACS
Packaging Poly Bottle
Specific Gravity 1.57
Chemical Name or Material Potassium Acetate, ACS Reagent
Product Title
Select an issue

By clicking Submit, you acknowledge that you may be contacted by Fisher Scientific in regards to the feedback you have provided in this form. We will not share your information for any other purposes. All contact information provided shall also be maintained in accordance with our Privacy Policy.