Promotional price valid on web orders only. Your contract pricing may differ. Interested in signing up for a dedicated account number?
Learn More

Potassium Acetate, BiotechGrade, 99-100.5%, Spectrum™ Chemical
SDP

Catalog No. 18605428 Shop All Spectrum Chemical Mfg. Corp. Products
Change view
Click to view available options
Quantity:
500 g
2.5 kg
Packaging:
Poly Bottle
2 product options available for selection
Product selection table with 2 available options. Use arrow keys to navigate and Enter or Space to select.
Catalog No. Quantity Packaging
18-605-428 2.5 kg Poly Bottle
18-605-427 500 g Poly Bottle
Use arrow keys to navigate between rows. Press Enter or Space to select a product option. 2 options available.
2 options
Catalog No. 18-605-428 Supplier Spectrum Chemical Manufacturing Corporation Supplier No. P11882.5KG
Get Up to $500 in Chemicals with Purchase
View all promotions
Add to Cart
Add to Cart

missing translation for 'validMainframeMsg'

P1188, 127-08-2, C2H3KO2

Spectrum™ Chemical Potassium Acetate, BiotechGrade is used in molecular biology to precipitate dodecyl sulfate (DS) and DS-bound proteins, allowing the removal of proteins from DNA. It is also used as a salt for the ethanol precipitation of DNA. Potassium acetate is also used in mixtures applied for tissue preservation, fixation, and mummification.

Chemical Identifiers

CAS 127-08-2
Molecular Formula C2H3KO2
Molecular Weight (g/mol) 98.14
InChI Key SCVFZCLFOSHCOH-UHFFFAOYSA-M
IUPAC Name potassium acetate
SMILES [K+].CC([O-])=O

Specifications

pH 7.5 to 8.5
CAS Max % 1
Quantity 2.5 kg
Percent Purity 99.0 to 100.5%
Grade Biotech
Loss on Drying 0.01
Odor Odorless
Packaging Poly Bottle
Appearance Colorless crystal
Product Title
Select an issue

By clicking Submit, you acknowledge that you may be contacted by Fisher Scientific in regards to the feedback you have provided in this form. We will not share your information for any other purposes. All contact information provided shall also be maintained in accordance with our Privacy Policy.