Learn More
Potassium Acetate, Crystalline Powder, USP, 99-100.5%, Spectrum™ Chemical
Shop All Spectrum Chemical Mfg. Corp. Products

Description
Specifications
Specifications
| pH | 7.5 to 8.5 |
| CAS Max % | 1 |
| Physical Form | Crystalline Powder, Needles or Crystals |
| Quantity | 50 kg |
| Percent Purity | 99 to 100.5% |
| Grade | USP |
| Loss on Drying | 0.01 |
| Packaging | Poly Bottle |
| Chemical Name or Material | Potassium Acetate, Crystalline Powder |
By clicking Submit, you acknowledge that you may be contacted by Fisher Scientific in regards to the feedback you have provided in this form. We will not share your information for any other purposes. All contact information provided shall also be maintained in accordance with our Privacy Policy.