Promotional price valid on web orders only. Your contract pricing may differ. Interested in signing up for a dedicated account number?
Learn More

Potassium Acetate Crystals, GR, ACS, MilliporeSigma™

Catalog No. MPX13303 Shop All MilliporeSigma Products
Change view
Click to view available options
Quantity:
2 kg
Packaging:
Plastic Bottle
1 product options available for selection
Product selection table with 1 available options. Use arrow keys to navigate and Enter or Space to select.
Catalog No. Quantity Packaging
MPX13303 2 kg Plastic Bottle
Use arrow keys to navigate between rows. Press Enter or Space to select a product option. 1 options available.
1 options
Catalog No. MPX13303 Supplier MilliporeSigma Supplier No. PX13303

Item Discontinued This item has been discontinued by the supplier. Please to view product availability in your area.

View Alternate Product

Chemical Identifiers

CAS 127-08-2
Molecular Formula C2H3KO2
Molecular Weight (g/mol) 98.142
InChI Key SCVFZCLFOSHCOH-UHFFFAOYSA-M
Synonym potassium acetate, acetic acid, potassium salt, diuretic salt, potassium ethanoate, koac, octan draselny czech, potassiumacetate, acetic acid potassium salt, acok, fema no. 2920
PubChem CID 517044
ChEBI CHEBI:32029
IUPAC Name potassium;acetate
SMILES CC(=O)[O-].[K+]

Specifications

Melting Point 292°C
Color White
pH 7.5 to 8.5 (50g/L, H2O, 20°C)
Physical Form Crystalline Powder and/or Crystals
Quantity 2 kg
Solubility Information 2530 g/L
Formula Weight 98.14
Percent Purity 99.0%
Grade Meets ACS Specifications, Meets Reagent Specifications for testing USP/NF monographs
Odor Weakly of acetic acid
Packaging Plastic Bottle
Density 1.57 g/cc at 25°C
Chemical Name or Material Potassium Acetate
Show More Show Less
EINECSNumber 204-822-2
Recommended Storage Store at room temperature
Product Title
Select an issue

By clicking Submit, you acknowledge that you may be contacted by Fisher Scientific in regards to the feedback you have provided in this form. We will not share your information for any other purposes. All contact information provided shall also be maintained in accordance with our Privacy Policy.