Learn More
HIMEDIA Potassium Acetate, Hi-AR/ACS, GRM3930, Laboratory Chemicals, Organic Salt, Analytical Reagent, ACS Grade, CAS 127-08-2, Buffer Reagent, Powder, 500 g
Catalog No. NC4013651
Encompass_Preferred
Potassium acetate, Hi-AR/ACS is an analytical reagent and ACS grade salt (C2H3KO2, CAS 127-08-2, molecular weight 98.14) supplied as colourless to white deliquescent crystals or powder. A 5% aqueous solution gives a pH of 6.50 to 9.00, with chloride held to 0.003% maximum and tight limits on heavy metals, supporting reliable results in buffer preparation, molecular biology precipitations, dehydrating applications and general analytical chemistry. Store below 30 C, shelf life four years. Packaged as 500 g.
Catalog No. NC4013651 Supplier HIMEDIA Supplier No. GRM3930500G
Potassium acetate, Hi-AR/ACS is an analytical reagent and ACS grade salt (C2H3KO2, CAS 127-08-2, molecular weight 98.14) supplied as colourless to white deliquescent crystals or powder. A 5% aqueous solution gives a pH of 6.50 to 9.00, with chloride held to 0.003% maximum and tight limits on heavy metals, supporting reliable results in buffer preparation, molecular biology precipitations, dehydrating applications and general analytical chemistry. Store below 30 C, shelf life four years. Packaged as 500 g.