Learn More
Medchemexpress LLC Potassium acetate, for molecular biology | 127-08-2 | MFCD00012458 | 99.0% | 98.14 g/mol | C2H3KO2 | 5g
SDP
Catalog No. 5000269771
Encompass_Preferred
Acetic acid potassium 99% is a potassium salt employed to replenish electrolytes for restoration of water-electrolyte balance Acetic acid potassium 99% can employ in DNA and protein purification Acetic acid potassium 99% has been used to prepare neutralizing solution for alkaline lysis of bacteria[1][2]
Catalog No. 50-002-69771 Supplier Medchemexpress LLC Supplier No. HYY0319B5G
Acetic acid potassium 99% is a potassium salt employed to replenish electrolytes for restoration of water-electrolyte balance Acetic acid potassium 99% can employ in DNA and protein purification Acetic acid potassium 99% has been used to prepare neutralizing solution for alkaline lysis of bacteria[1][2]