Promotional price valid on web orders only. Your contract pricing may differ. Interested in signing up for a dedicated account number?
Learn More

Potassium Acid Phthalate, 0.0500 N (N/20), Ricca Chemical
SDP

Catalog No. 587032 Shop All Ricca Chemical Company Products
Change view
Click to view available options
Quantity:
500 mL
1 L
4 L
3 product options available for selection
Product selection table with 3 available options. Use arrow keys to navigate and Enter or Space to select.
Catalog No. Quantity
5870-32 1 L
5870-16 500 mL
58701 4 L
Use arrow keys to navigate between rows. Press Enter or Space to select a product option. 3 options available.
3 options
Catalog No. 5870-32 Supplier Ricca Chemical Company Supplier No. 587032
Add to Cart
Add to Cart

missing translation for 'validMainframeMsg'

For Precision Titrations Testing

Specifications

Chemical Name or Material Potassium Acid Phthalate
CAS 7732-18-5
Grade Laboratory
Packaging Natural Poly Bottle
Methods APHA for Acidity
pH ∼4
Quantity 1 L
Product Title
Select an issue

By clicking Submit, you acknowledge that you may be contacted by Fisher Scientific in regards to the feedback you have provided in this form. We will not share your information for any other purposes. All contact information provided shall also be maintained in accordance with our Privacy Policy.