Learn More
Hach Company Potassium Acid Phthalate (KHP), 500 g
Catalog No. 501994245 Shop All Hach Company Products
Encompass
Primary standard acid, ACS Grade, CAS No. 877-24-7 C8H5O4K, FW = 204.23 Assay = ≥99.95 but ≤100.05%
Catalog No. 50-199-4245 Supplier Hach Company Supplier No. 31534
Primary standard acid, ACS Grade, CAS No. 877-24-7 C8H5O4K, FW = 204.23 Assay = ≥99.95 but ≤100.05%