Promotional price valid on web orders only. Your contract pricing may differ. Interested in signing up for a dedicated account number?
Learn More

Potassium Acid Phthalate, ACS Reagent, Primary Standard (Potassium Hydrogen Phthalate), Reagents

Catalog No. 01337874
Change view
Click to view available options
Quantity:
125 g
250 g
500 g
3 product options available for selection
Product selection table with 3 available options. Use arrow keys to navigate and Enter or Space to select.
Catalog No. Quantity
01-337-874 500 g
01-337-876 125 g
01-337-875 250 g
Use arrow keys to navigate between rows. Press Enter or Space to select a product option. 3 options available.
3 options
Catalog No. 01-337-874 Supplier Reagents Holdings Llc Supplier No. C1276500500A5
Add to Cart
Add to Cart

Potassium Acid Phthalate, ACS Reagent, Primary Standard (Potassium Hydrogen Phthalate), ACS, for general laboratory use. Manufactured in ISO 9001 facility.

Specifications

Chemical Name or Material Potassium Acid Phthalate, ACS Reagent, Primary Standard (Potassium Hydrogen Phthalate)
CAS 877-24-7
Melting Point 0°C
Boiling Point 378°C
Color White
Molecular Formula HOCOC6H4COOK
Synonym Potassium Hydrogen Phthalate, KHP, Potassium Biphthalate
Molecular Weight (g/mol) 204.22
Grade ACS
Quantity 500 g
Packaging Poly Bottle
Physical Form Solid
Specific Gravity 1.636
Show More Show Less
Product Title
Select an issue

By clicking Submit, you acknowledge that you may be contacted by Fisher Scientific in regards to the feedback you have provided in this form. We will not share your information for any other purposes. All contact information provided shall also be maintained in accordance with our Privacy Policy.