Promotional price valid on web orders only. Your contract pricing may differ. Interested in signing up for a dedicated account number?
Learn More

Sigma Aldrich Potassium Antimony(III) Tartrate Hydrate

Catalog No. 244791500G Shop All MilliporeSigma Sigma Organic Chemistry Products
Encompass_Preferred
Change view
Click to view available options
Quantity:
100 g
500 g
2 product options available for selection
Product selection table with 2 available options. Use arrow keys to navigate and Enter or Space to select.
Catalog No. Quantity
24-479-1500G 500 g
24-479-1100G 100 g
Use arrow keys to navigate between rows. Press Enter or Space to select a product option. 2 options available.
2 options
Danger
Catalog No. 24-479-1500G Supplier Sigma Aldrich Supplier No. 244791500G
  • NEW LOWER PRICE
May include imposed supplier surcharges.
Add to Cart
Add to Cart

missing translation for 'validMainframeMsg'

Danger

MilliporeSigma Sigma Organics products encompass a wide range of quality reagents, solvents, catalysts, and building blocks for organic synthesis. From benchtop discovery to process development and scale-up, Sigma Organics solutions are built to meet the needs of synthetic chemists.

Potassium antimony(III) tartrate hydrate may be used as a standard during the quantification of Sb species present in the juices by HPLC. It may also be used in the preparation of novel Sb(III)-containing polyanions by reacting with trilacunary Keggin-type polyanions.

Specifications

CAS 331753-56-1
Density 2.6 g/mL at 25°C
Molecular Formula C8H4K2O12Sb2 -+ xH2O
Linear Formula C8H4K2O12Sb2 -+ xH2O
MDL Number MFCD00149437
Quantity 500 g
Synonym Antimony potassium tartrate
Molecular Weight (g/mol) 613.83 g/mol
Product Title
Select an issue

By clicking Submit, you acknowledge that you may be contacted by Fisher Scientific in regards to the feedback you have provided in this form. We will not share your information for any other purposes. All contact information provided shall also be maintained in accordance with our Privacy Policy.