Promotional price valid on web orders only. Your contract pricing may differ. Interested in signing up for a dedicated account number?
Learn More

Sigma Aldrich Potassium antimony(III) tartrate hydrate

Catalog No. 23005710G Shop All MilliporeSigma Sigma Organic Chemistry Products
Encompass_Preferred
Change view
Click to view available options
Quantity:
10 g
1 product options available for selection
Product selection table with 1 available options. Use arrow keys to navigate and Enter or Space to select.
Catalog No. Quantity
23-005-710G 10 g
Use arrow keys to navigate between rows. Press Enter or Space to select a product option. 1 options available.
1 options
Catalog No. 23-005-710G Supplier Sigma Aldrich Supplier No. 23005710G
  • NEW LOWER PRICE
May include imposed supplier surcharges.
Add to Cart
Add to Cart

missing translation for 'validMainframeMsg'

MilliporeSigma Sigma Organics products encompass a wide range of quality reagents, solvents, catalysts, and building blocks for organic synthesis. From benchtop discovery to process development and scale-up, Sigma Organics solutions are built to meet the needs of synthetic chemists.

Potassium antimony(III) tartrate hydrate

Specifications

CAS 331753-56-1
Density 2.6 g/mL (at 25°C)
Molecular Formula C8H4K2O12Sb2 -+ xH2O
Linear Formula C8H4K2O12Sb2 -+ xH2O
Quantity 10 g
MDL Number MFCD00149437
Synonym Antimony potassium tartrate
Molecular Weight (g/mol) 613.83
Percent Purity 99.95% trace metals basis
Product Title
Select an issue

By clicking Submit, you acknowledge that you may be contacted by Fisher Scientific in regards to the feedback you have provided in this form. We will not share your information for any other purposes. All contact information provided shall also be maintained in accordance with our Privacy Policy.