Promotional price valid on web orders only. Your contract pricing may differ. Interested in signing up for a dedicated account number?
Learn More

Potassium Antimony Tartrate Solution, For Phosphorus, LabChem™

Catalog No. LC187201 Shop All LabChem Products
Change view
Click to view available options
Quantity:
500 mL
1 L
2 product options available for selection
Product selection table with 2 available options. Use arrow keys to navigate and Enter or Space to select.
Catalog No. Quantity
LC187201 500 mL
LC187202 1 L
Use arrow keys to navigate between rows. Press Enter or Space to select a product option. 2 options available.
2 options
Catalog No. LC187201 Supplier LabChem, Inc. Supplier No. LC187201
Add to Cart
Add to Cart

missing translation for 'validMainframeMsg'

Specifications

Chemical Name or Material Potassium Antimony Tartrate Solution
Color Colorless
Name Note For Phosphorus
CAS Max % 99.23, 0.495, 0.27, 0.005
CAS 67-66-3, 28300-74-5, 64-17-5
Linear Formula C8H4K2O12Sb2·3H2O
Synonym water, dihydrogen oxide, distilled water, purified water, water vapor, sterile water, water, purified, water, deionized, water, mineral, dihydrogen monoxide
Solubility Information Soluble in water
Formula Weight 667.87
Identification Passes Test
Packaging Poly Bottle
Decomposition Information Antimony and its oxides; Carbon monoxide; Carbon dioxide
Quantity 500 mL
Physical Form Liquid
Show More Show Less
WARNING: Cancer and Reproductive Harm - www.P65Warnings.ca.gov
Product Title
Select an issue

By clicking Submit, you acknowledge that you may be contacted by Fisher Scientific in regards to the feedback you have provided in this form. We will not share your information for any other purposes. All contact information provided shall also be maintained in accordance with our Privacy Policy.