Promotional price valid on web orders only. Your contract pricing may differ. Interested in signing up for a dedicated account number?
Learn More

Sigma Aldrich Potassium aquapentachlororuthenate(III)

Catalog No. 2837381G Shop All MilliporeSigma Sigma Organic Chemistry Products
Encompass
Encompass_Preferred
Change view
Click to view available options
Quantity:
1 g
1 product options available for selection
Product selection table with 1 available options. Use arrow keys to navigate and Enter or Space to select.
Catalog No. Quantity
28-373-81G 1 g
Use arrow keys to navigate between rows. Press Enter or Space to select a product option. 1 options available.
1 options
Catalog No. 28-373-81G Supplier Sigma Aldrich Supplier No. 2837381G
May include imposed supplier surcharges.
Add to Cart
Add to Cart

MilliporeSigma Sigma Organics products encompass a wide range of quality reagents, solvents, catalysts, and building blocks for organic synthesis. From benchtop discovery to process development and scale-up, Sigma Organics solutions are built to meet the needs of synthetic chemists.

The product may be used for eletrodeposition of bifunctional catalyst films, PtRu on nanoporous TiO2 substrates.

Specifications

CAS 14404-33-2
Molecular Formula H2Cl5K2ORu
Linear Formula K2RuCl5(H2O)
MDL Number MFCD00011524
Quantity 1 g
Synonym Dipotassium aquapentachlororuthenate(III)
Molecular Weight (g/mol) 374.55
Percent Purity 26-30%Ru basis (gravimetric)
Biological Activity Respiratory system
Product Title
Select an issue

By clicking Submit, you acknowledge that you may be contacted by Fisher Scientific in regards to the feedback you have provided in this form. We will not share your information for any other purposes. All contact information provided shall also be maintained in accordance with our Privacy Policy.