Promotional price valid on web orders only. Your contract pricing may differ. Interested in signing up for a dedicated account number?
Learn More

Potassium aquapentachlororuthenate(III), Premion™, 99.99% (metals basis), Ru 26.4% min

Catalog No. AA4171603
Change view
Click to view available options
Quantity:
1 g
5 g
2 product options available for selection
Product selection table with 2 available options. Use arrow keys to navigate and Enter or Space to select.
Catalog No. Quantity
AA4171603 1 g
AA4171606 5 g
Use arrow keys to navigate between rows. Press Enter or Space to select a product option. 2 options available.
2 options
Catalog No. AA4171603 Supplier Thermo Scientific Chemicals Supplier No. 04171603
Add to Cart

CAS: 14404-33-2 | Cl5K2Ru | 356.52 g/mol

Potassium aquapentachlororuthenate(III) is used in Hydrogenation (Slurry Bed).It is also used as an intermediates.

This Thermo Scientific Chemicals brand product was originally part of the Alfa Aesar product portfolio. Some documentation and label information may refer to the legacy brand. The original Alfa Aesar product / item code or SKU reference has not changed as a part of the brand transition to Thermo Scientific Chemicals.

TRUSTED_SUSTAINABILITY

Chemical Identifiers

CAS 14404-33-2
Molecular Formula Cl5K2Ru
Molecular Weight (g/mol) 356.52
MDL Number MFCD00058747
InChI Key IXYMQZXAJLWXRK-UHFFFAOYSA-I
Synonym dipotassium aquapentachlororuthenate, potassium pentachlororuthenate iii hydrate, dipotassium aquapentachlororuthenate iii, dipotassium pentachlororutheniumdiuide hydrate, potassium aquapentachlororuthenate iii , premion, ruthenate 2-, aquapentachloro-, dipotassium, oc-6-21
PubChem CID 16211344
IUPAC Name dipotassium;pentachlororuthenium(2-);hydrate
SMILES [K+].[K+].Cl[Ru--](Cl)(Cl)(Cl)Cl

Specifications

Color Brown to Red
Physical Form Crystalline
Quantity 1 g
Solubility Information Soluble in water.
Formula Weight 374.55 (356.53 Anhydrous)
Percent Purity 99.99%
Grade Premion
Concentration or Composition (by Analyte or Components) Ru 26.4% min.
Assay (metals basis)
Chemical Name or Material Potassium aquapentachlororuthenate(III)
Hazard Category Warning
Hazard Statement GHS H Statement
H315-H319-H335
Causes skin irritation.
Causes serious eye irritation.
May cause respiratory irritation.
Precautionary Statement GHS P Statement
P261-P280-P305+P351+P338-P304+P340-P405-P501a
Avoid breathing dust/fume/gas/mist/vapors/spray.
Wear protective gloves/protective clothing/eye protection/face protection.
IF IN EYES: Rinse cautiously with water for several minutes.
Remove contact lenses, if present and easy to do.
Continue rinsing.
IF INHALED: Remove to fresh air and keep at rest in a position comfortable for breathing.
Store locked up.
Dispose of contents/container in accordance with local/regional/national/international regulations.
EINECSNumber 238-374-4
TSCA No
Recommended Storage Ambient temperatures

RUO – Research Use Only

Product Title
Select an issue

By clicking Submit, you acknowledge that you may be contacted by Fisher Scientific in regards to the feedback you have provided in this form. We will not share your information for any other purposes. All contact information provided shall also be maintained in accordance with our Privacy Policy.