Learn More
Crystal Chem Inc Potassium Assay Kit, 96-tests
SDP
Catalog No. 501947669
Encompass_Preferred
The Potassium Assay Kit is used to quantify levels of potassium in serum using only a 5 µL sample volume. Highlights: Liquid stable format requires no reagent preparation and reduces waste. Results in as little as ten minutes. Convenient and cost effective. Crystal Chem is a worldwide leading supplier of ELISA kits for human markers. Our kits are among the highest quality found in the market, and are used by leading research universities, pharma companies, and government institutions around the world.
Catalog No. 50-194-7669 Supplier Crystal Chem Inc Supplier No. 80169
The Potassium Assay Kit is used to quantify levels of potassium in serum using only a 5 µL sample volume. Highlights: Liquid stable format requires no reagent preparation and reduces waste. Results in as little as ten minutes. Convenient and cost effective. Crystal Chem is a worldwide leading supplier of ELISA kits for human markers. Our kits are among the highest quality found in the market, and are used by leading research universities, pharma companies, and government institutions around the world.