Promotional price valid on web orders only. Your contract pricing may differ. Interested in signing up for a dedicated account number?
Learn More

Potassium Benzoate, FCC, 99-101%, Spectrum™ Chemical
SDP

Catalog No. 18605445 Shop All Spectrum Chemical Manufacturing Corporation Products
Change view
Click to view available options
Quantity:
500 g
Packaging:
Poly Bottle
1 product options available for selection
Product selection table with 1 available options. Use arrow keys to navigate and Enter or Space to select.
Catalog No. Quantity Packaging
18-605-445 500 g Poly Bottle
Use arrow keys to navigate between rows. Press Enter or Space to select a product option. 1 options available.
1 options
Catalog No. 18-605-445 Supplier Spectrum Chemical Manufacturing Corporation Supplier No. PO156500GM

PO156, 582-25-2, C7H5KO2

Spectrum™ Chemical Potassium Benzoate, FCC is used as a preservative that limits the growth of yeast, mold and some bacteria. The FCC grade meets the requirements of the Food Chemical Codex indicates and is suitable for all food, beverage and nutritional supplement applications. Spectrum Chemical offers over 300 Food grade chemical ingredients packaged in laboratory size bottles to production drum quantities and are manufactured, packaged and stored under current Good Manufacturing Practices (cGMP) per 21CFR part 211 in FDA registered and inspected facilities.

Chemical Identifiers

CAS 582-25-2
Molecular Formula C7H5KO2
Molecular Weight (g/mol) 160.21
InChI Key XAEFZNCEHLXOMS-UHFFFAOYSA-M
IUPAC Name potassium benzoate
SMILES [K+].[O-]C(=O)C1=CC=CC=C1

Specifications

CAS Max % 1
Packaging Poly Bottle
Quantity 500 g
Percent Purity 99 to 101%
Grade FCC
Product Title
Select an issue

By clicking Submit, you acknowledge that you may be contacted by Fisher Scientific in regards to the feedback you have provided in this form. We will not share your information for any other purposes. All contact information provided shall also be maintained in accordance with our Privacy Policy.