Promotional price valid on web orders only. Your contract pricing may differ. Interested in signing up for a dedicated account number?
Learn More

Potassium Bicarbonate, 99.7-100.5%, (ACS Reagent Grade), MP Biomedicals

Catalog No. ICN15255780 Shop All MP Biomedicals, Inc Products
Change view
Click to view available options
Quantity:
100 g
1 kg
5 kg
3 product options available for selection
Product selection table with 3 available options. Use arrow keys to navigate and Enter or Space to select.
Catalog No. Quantity
ICN15255780 100 g
ICN15255794 5 kg
ICN15255791 1 kg
Use arrow keys to navigate between rows. Press Enter or Space to select a product option. 3 options available.
3 options
Catalog No. ICN15255780 Supplier MP Biomedicals, Inc Supplier No. 0215255780
Add to Cart
Add to Cart

Chemical Identifiers

CAS 298-14-6
Molecular Formula CHKO3
Molecular Weight (g/mol) 100.11
MDL Number MFCD00011402
InChI Key TYJJADVDDVDEDZ-UHFFFAOYSA-M
Synonym potassium bicarbonate, potassium hydrogen carbonate, potassium hydrogencarbonate, potassium acid carbonate, monopotassium carbonate, carbonic acid, monopotassium salt, unii-hm5z15lebn, potassiumbicarbonate, ccris 3510, khco3
PubChem CID 516893
ChEBI CHEBI:81862
IUPAC Name potassium hydrogen carbonate
SMILES [K+].OC([O-])=O

Specifications

Quantity 100 g
Assay Percent Range 99.7-100.5%
Formula Weight 100.12
Grade ACS Reagent
Chemical Name or Material Potassium Bicarbonate
Recommended Storage Room Temperature
Product Title
Select an issue

By clicking Submit, you acknowledge that you may be contacted by Fisher Scientific in regards to the feedback you have provided in this form. We will not share your information for any other purposes. All contact information provided shall also be maintained in accordance with our Privacy Policy.