Learn More
Sigma Aldrich Fine Chemicals Biosciences Potassium bicarbonate 99.950G
Catalog No. 502899471
Encompass
Potassium bicarbonate is water soluble alkaline potassium salt with monoclinic crystalline structure. It is a raw material for the synthesis of many potassium compounds. It is a better coolant than sodium bicarbonate in the aerosol fire extinguishing apparatus. It shows potential as an antifungal agent.
Catalog No. 50-289-9471 Supplier Sigma Aldrich Fine Chemicals Biosciences Supplier No. 43158350G
Potassium bicarbonate is water soluble alkaline potassium salt with monoclinic crystalline structure. It is a raw material for the synthesis of many potassium compounds. It is a better coolant than sodium bicarbonate in the aerosol fire extinguishing apparatus. It shows potential as an antifungal agent.