Promotional price valid on web orders only. Your contract pricing may differ. Interested in signing up for a dedicated account number?
Learn More

Potassium Bicarbonate, Purified (Potassium Hydrogen Carbonate), Reagents

Catalog No. 01337124
Change view
Click to view available options
Quantity:
100 g
500 g
2 product options available for selection
Product selection table with 2 available options. Use arrow keys to navigate and Enter or Space to select.
Catalog No. Quantity
01-337-124 500 g
01-337-125 100 g
Use arrow keys to navigate between rows. Press Enter or Space to select a product option. 2 options available.
2 options
Catalog No. 01-337-124 Supplier Reagents Holdings Llc Supplier No. C2277000500A5
Add to Cart
Add to Cart

missing translation for 'validMainframeMsg'

Potassium Bicarbonate, Purified (Potassium Hydrogen Carbonate), Chemically Pure, for general laboratory use. Manufactured in ISO 9001 facility.

Chemical Identifiers

CAS 298-14-6
Molecular Formula CHKO3
Molecular Weight (g/mol) 100.11
MDL Number MFCD00011402
InChI Key TYJJADVDDVDEDZ-UHFFFAOYSA-M
Synonym Potassium Hydrogen Carbonate
IUPAC Name potassium hydrogen carbonate
SMILES [K+].OC([O-])=O

Specifications

Melting Point 127°C
Color Colorless/White
Physical Form Solid
Quantity 500 g
Grade Pure
Packaging Poly Bottle
Specific Gravity 2.17
Chemical Name or Material Potassium Bicarbonate, Purified (Potassium Hydrogen Carbonate)
Product Title
Select an issue

By clicking Submit, you acknowledge that you may be contacted by Fisher Scientific in regards to the feedback you have provided in this form. We will not share your information for any other purposes. All contact information provided shall also be maintained in accordance with our Privacy Policy.