Promotional price valid on web orders only. Your contract pricing may differ. Interested in signing up for a dedicated account number?
Learn More

Potassium bicarbonate, Puriss., meets analytical spec. of BP, Ph. Eur., USP, E501, 99.5-101.0% (acidimetric), Solstice

Catalog No. 60044737 Shop All Solstice Research Chemicals Products
Change view
Click to view available options
Quantity:
1 kg
6 x 1 kg
Packaging:
Poly Bottle
2 product options available for selection
Product selection table with 2 available options. Use arrow keys to navigate and Enter or Space to select.
Catalog No. Quantity Packaging
60-044-737 6 x 1 kg Poly Bottle
60-019-39 1 kg Poly Bottle
Use arrow keys to navigate between rows. Press Enter or Space to select a product option. 2 options available.
2 options
Catalog No. 60-044-737 Supplier Solstice Supplier No. 126026X1KG
Add to Cart
Add to Cart

Please call Customer Service at 1-800-234-7437 or send an email to help@thermofisher.com for assistance.

Puriss., meets analytical specification of BP, Ph. Eur., USP, E501, 99.5-101.0% (acidimetric)

  • Leading the Industry For Over 65 Years
  • Solstice now delivers premium grade inorganic reagents worldwide – with consistency, purity, and accuracy assured
TRUSTED_SUSTAINABILITY

Chemical Identifiers

CAS 298-14-6
Molecular Formula CHKO3
Molecular Weight (g/mol) 100.11
MDL Number MFCD00011402
InChI Key TYJJADVDDVDEDZ-UHFFFAOYSA-M
Synonym potassium bicarbonate, potassium hydrogen carbonate, potassium hydrogencarbonate, potassium acid carbonate, monopotassium carbonate, carbonic acid, monopotassium salt, unii-hm5z15lebn, potassiumbicarbonate, ccris 3510, khco3
PubChem CID 516893
ChEBI CHEBI:81862
IUPAC Name potassium;hydrogen carbonate
SMILES [K+].OC([O-])=O

Specifications

CAS Max % 100.0000%
Physical Form Powder
Quantity 6 x 1 kg
Assay Percent Range 99.5 to 101.0%
Linear Formula KHCO3
UN Number NONH for all modes of transport
Beilstein 4535309
Solubility Information Soluble in water
Formula Weight 100.12
Percent Purity 99.5 to 101.0% (acidimetric)
Grade puriss.
Packaging Poly Bottle
Density 2.17 g/cm3
Chemical Name or Material Potassium bicarbonate, Puriss., meets analytical specification of BP, Ph. Eur., USP, E501, 99.5-101.0% (acidimetric)
Show More Show Less
ShelfLife 1650 days from date of manufacture
EINECSNumber 206-059-0
Recommended Storage Room Temp
WARNING: Reproductive Harm - www.P65Warnings.ca.gov
Product Title
Select an issue

By clicking Submit, you acknowledge that you may be contacted by Fisher Scientific in regards to the feedback you have provided in this form. We will not share your information for any other purposes. All contact information provided shall also be maintained in accordance with our Privacy Policy.