Learn More
Hach Company Potassium Biiodate Standard Solution, 0.025 N, 500 mL
Supplier:  Hach Company 31249H
Encompass
Standard oxidant solution, for standardization of thiosulfate and phenylarsine oxide titrants
Catalog No. 50-199-7436