Promotional price valid on web orders only. Your contract pricing may differ. Interested in signing up for a dedicated account number?
Learn More

Potassium Biiodate, 0.0282 N (0.00235 M), 1000 ppm as Cl2, Ricca Chemical
SDP

Catalog No. 587716 Shop All Ricca Chemical Company Products
Change view
Click to view available options
Quantity:
500 mL
1 L
4 L
3 product options available for selection
Product selection table with 3 available options. Use arrow keys to navigate and Enter or Space to select.
Catalog No. Quantity
587716 500 mL
5877-32 1 L
5877-1 4 L
Use arrow keys to navigate between rows. Press Enter or Space to select a product option. 3 options available.
3 options
Catalog No. 587716 Supplier Ricca Chemical Company Supplier No. 587716
Add to Cart
Add to Cart

missing translation for 'validMainframeMsg'

1 mL = 1 mg Cl2, ASTM for Chlorine Testing

Specifications

Chemical Name or Material Potassium Biiodate
Color Colorless
CAS 13455-24-8, 7732-18-5
CAS Min % 98.0, 0.09
CAS Max % 100.0, 0.09
Molecular Formula HI2KO6
Synonym potassium hydrogen diiodate, potassium biiodate, kaliumhydrogendijodat, iodic acid hio3 , potassium salt 2:1, potassium biiodate solution, potassium iodic acid iodate, io3.k.hio3, potassium ion iodic acid iodate, potassium hydrogen iodate powder, potassium hydrogen diiodate, p.a
InChI Key ACAYDTMSDROWHW-UHFFFAOYSA-M
SMILES OI(=O)=O.[O-]I(=O)=O.[K+]
IUPAC Name potassium;iodic acid;iodate
Molecular Weight (g/mol) 389.909
PubChem CID 23700942
Equivalence 1 mL = 1 mg Cl
Grade Laboratory
Packaging Amber Glass Bottle
Quantity 500 mL
Physical Form Liquid
Concentration or Composition Notes 0.0282Normal,0.00235Molar
Show More Show Less
Product Title
Select an issue

By clicking Submit, you acknowledge that you may be contacted by Fisher Scientific in regards to the feedback you have provided in this form. We will not share your information for any other purposes. All contact information provided shall also be maintained in accordance with our Privacy Policy.