Promotional price valid on web orders only. Your contract pricing may differ. Interested in signing up for a dedicated account number?
Learn More

Potassium Bis(1-pyrazolyl)borohydride 98.0+%, TCI America™
SDP

Catalog No. P14395G Shop All Tokyo Chemical Industry (TCI) Products
Change view
Click to view available options
Quantity:
1 g
5 g
2 product options available for selection
Product selection table with 2 available options. Use arrow keys to navigate and Enter or Space to select.
Catalog No. Quantity
P14395G 5 g
P14391G 1 g
Use arrow keys to navigate between rows. Press Enter or Space to select a product option. 2 options available.
2 options
Catalog No. P14395G Supplier TCI America Supplier No. P14395G
Add to Cart
Add to Cart

missing translation for 'validMainframeMsg'

Chemical Identifiers

CAS 18583-59-0
Molecular Formula C6H6BKN4
Molecular Weight (g/mol) 184.05
MDL Number MFCD00058739
InChI Key KIXPNQZZOUCWGN-UHFFFAOYSA-N
Synonym Potassium Dihydrobis(1-pyrazolyl)borate
PubChem CID 44630456
IUPAC Name potassium;di(pyrazol-1-yl)boranide
SMILES [B-](N1C=CC=N1)N2C=CC=N2.[K+]

Specifications

Melting Point 168°C
Color White
Quantity 5 g
UN Number 2813
Formula Weight 186.07
Percent Purity ≥98.0% (T)
Physical Form Crystalline Powder
Chemical Name or Material Potassium Bis(1-pyrazolyl)borohydride
Product Title
Select an issue

By clicking Submit, you acknowledge that you may be contacted by Fisher Scientific in regards to the feedback you have provided in this form. We will not share your information for any other purposes. All contact information provided shall also be maintained in accordance with our Privacy Policy.