Promotional price valid on web orders only. Your contract pricing may differ. Interested in signing up for a dedicated account number?
Learn More
Learn More
Sigma Aldrich Potassium Bis(Trimethylsilyl)Amide Solution
Shop All MilliporeSigma Sigma Organic Chemistry Products
Description
We are committed to bringing you Greener Alternative Products, which adhere to one or more of The 12 Principles of Greener Chemistry. This product, 1.0 M in 2-methyltetrahydrofuran aligns with Safer Solvents and Auxiliaries, Use of Renewable Feedstocks and Inherently Safer Chemistry for Accident Prevention. Click here for more information.Strong organic base in greener solvent, 2-methyltetrahydrofuran (2-MeTHF).2-Methyltetrahydrofuran (2-MeTHF): A Biomass-Derived Solvent with Broad Application in Organic ChemistrySure/Seal is a trademark of Sigma-Aldrich Co. LLC
Specifications
Specifications
| CAS | 536724-67-1 |
| Melting Point | 159 (decomposition)°C to 162°C |
| Molecular Formula | C54H69Cl2N2PRu |
| Linear Formula | C54H69Cl2N2PRu |
| MDL Number | MFCD08667892 |
| Quantity | 500 mg |
| Synonym | Hexamethyldisilazane potassium salt solution; Potassium hexamethyldisilazide solution |
| Molecular Weight (g/mol) | 949.09 g/mol |
missing translation for 'productTitle'
missing translation for 'byClickingSubmit' missing translation for 'privacyPolicy'.
missing translation for 'spotOppurtunityImprovement'