Learn More
Medchemexpress LLC Potassium bisperoxo(1,10-phenanthroline) oxovanadate (V) | 42494-73-5 | 98.0% | C12H8KN2O5V | 10MG
SDP
Supplier:  Medchemexpress LLC HY13606510MG
Encompass_Preferred
bpV(phen) is a vanadium-based small-molecule inhibitor of protein tyrosine phosphatases, including PTEN. As an insulin-mimetic agent, it is used in cellular and biochemical studies to probe insulin signaling, apoptosis, angiogenesis, and parasite biology.
  • Potent PTEN inhibition with IC50 ≈ 38 nM and activity against other PTPs.
  • Insulin-mimetic activity for studies of insulin signaling pathways.
  • Useful for apoptosis, anti-angiogenesis, and anti-parasitic research applications.
  • High purity (98.0%) for reliable experimental results.
  • Solid, light yellow to yellow appearance and stable when stored as recommended.
Catalog No. 50-002-61938