Promotional price valid on web orders only. Your contract pricing may differ. Interested in signing up for a dedicated account number?
Learn More

Potassium bisulfate, 99%, extra pure

Catalog No. AC447610025
Change view
Click to view available options
Quantity:
500 g
2.5 kg
Packaging:
Glass bottle
2 product options available for selection
Product selection table with 2 available options. Use arrow keys to navigate and Enter or Space to select.
Catalog No. Quantity Packaging
AC447610025 2.5 kg Glass bottle
AC447615000 500 g Glass bottle
Use arrow keys to navigate between rows. Press Enter or Space to select a product option. 2 options available.
2 options
Catalog No. AC447610025 Supplier Thermo Scientific Chemicals Supplier No. 447610025
Add to Cart

Potassium bisulfate, 99%, HKO4S, CAS Number-7646-93-7, potassium bisulfate, sulfuric acid, monopotassium salt, sal enixum, potassium hydrogensulphate, potassium bisulphate, acid potassium sulfate, potassium acid sulfate, potassium hydrogen sulfate, monopotassium sulfate, caswell no. 682c, 500g | CAS: 7646-93-7 | HKO4S | 136.16 g/mol

This Thermo Scientific Chemicals brand product was originally part of the Acros Organics product portfolio. Some documentation and label information may refer to the legacy brand. The original Acros Organics product / item code or SKU reference has not changed as a part of the brand transition to Thermo Scientific Chemicals.

TRUSTED_SUSTAINABILITY

Chemical Identifiers

CAS 7646-93-7
Molecular Formula HKO4S
Molecular Weight (g/mol) 136.16
MDL Number MFCD00011404
InChI Key CHKVPAROMQMJNQ-UHFFFAOYSA-M
Synonym potassium bisulfate, potassium hydrogen sulfate, potassium bisulphate, sulfuric acid, monopotassium salt, potassium acid sulfate, sal enixum, potassium hydrogensulphate, monopotassium sulfate, acid potassium sulfate, caswell no. 682c
PubChem CID 516920
SMILES [K+].OS([O-])(=O)=O

Specifications

Melting Point 214.0°C
Quantity 2.5 kg
Assay Percent Range 99%
Linear Formula KHSO4
Merck Index 15,7734
Solubility Information (10% in water) Clear colorless
IUPAC Name potassium;hydrogen sulfate
Formula Weight 136.16
Percent Purity 99%
Grade Extra Pure
Packaging Glass bottle
Chemical Name or Material Potassium bisulfate
Show More Show Less
Hazard Category GHS Signal Word: Danger
Hazard Statement GHS H Statement
Causes severe skin burns and eye damage.
May cause respiratory irritation.
Precautionary Statement GHS P Statement
Immediately call a POISON CENTER or doctor/physician.
IF SWALLOWED: rinse mouth.
Do NOT induce vomiting.
IF ON SKIN (or hair): Take off immediately all contaminated clothing.
Rinse skin with water/shower.
IF IN
EINECSNumber 231-594-1

RUO – Research Use Only

Product Title
Select an issue

By clicking Submit, you acknowledge that you may be contacted by Fisher Scientific in regards to the feedback you have provided in this form. We will not share your information for any other purposes. All contact information provided shall also be maintained in accordance with our Privacy Policy.