Promotional price valid on web orders only. Your contract pricing may differ. Interested in signing up for a dedicated account number?
Learn More

Potassium Bisulfate (Crystalline/Certified), Fisher Chemical

Catalog No. p193500 Shop All Fisherbrand Products
Change view
Click to view available options
Quantity:
500 g
Packaging:
Glass Bottle
1 product options available for selection
Product selection table with 1 available options. Use arrow keys to navigate and Enter or Space to select.
Catalog No. Quantity Packaging
P193-500 500 g Glass Bottle
Use arrow keys to navigate between rows. Press Enter or Space to select a product option. 1 options available.
1 options
This item is not returnable. View return policy
Catalog No. P193-500
This item is not returnable. View return policy

Chemical Identifiers

CAS 7646-93-7
Molecular Formula HKO4S
Molecular Weight (g/mol) 136.16
MDL Number MFCD00011404
InChI Key CHKVPAROMQMJNQ-UHFFFAOYSA-M
Synonym potassium bisulfate, potassium hydrogen sulfate, potassium bisulphate, sulfuric acid, monopotassium salt, potassium acid sulfate, sal enixum, potassium hydrogensulphate, monopotassium sulfate, acid potassium sulfate, caswell no. 682c
PubChem CID 516920
IUPAC Name potassium hydrogen sulfate
SMILES [K+].OS([O-])(=O)=O

Specifications

Melting Point 214°C
Color White
pH 1
Physical Form Solid
Quantity 500 g
UN Number 2509
Formula Weight 136.17
Grade Certified
Identification Pass Test
Packaging Glass Bottle
Density 2.24g/cm³; Vapor Density: 10.0g/mol
Chemical Name or Material Potassium Bisulfate
Show More Show Less
Product Title
Select an issue

By clicking Submit, you acknowledge that you may be contacted by Fisher Scientific in regards to the feedback you have provided in this form. We will not share your information for any other purposes. All contact information provided shall also be maintained in accordance with our Privacy Policy.