Promotional price valid on web orders only. Your contract pricing may differ. Interested in signing up for a dedicated account number?
Learn More

Potassium Bisulfate, TraceSELECT™, ≥99.995% (metals basis), Solstice

Catalog No. 60047173 Shop All Solstice Research Chemicals Products
Change view
Click to view available options
Quantity:
25 g
Packaging:
Poly bottle
1 product options available for selection
Product selection table with 1 available options. Use arrow keys to navigate and Enter or Space to select.
Catalog No. Quantity Packaging
60-047-173 25 g Poly bottle
Use arrow keys to navigate between rows. Press Enter or Space to select a product option. 1 options available.
1 options
Catalog No. 60-047-173 Supplier Solstice Supplier No. 6034825G

missing translation for 'validMainframeMsg'

Chemical Identifiers

CAS 7646-93-7
Molecular Formula HKO4S
Molecular Weight (g/mol) 136.16
MDL Number MFCD00011404
InChI Key CHKVPAROMQMJNQ-UHFFFAOYSA-M
Synonym potassium bisulfate, potassium hydrogen sulfate, potassium bisulphate, sulfuric acid, monopotassium salt, potassium acid sulfate, sal enixum, potassium hydrogensulphate, monopotassium sulfate, acid potassium sulfate, caswell no. 682c
PubChem CID 516920
IUPAC Name potassium hydrogen sulfate
SMILES [K+].OS([O-])(=O)=O

Specifications

Melting Point ∼200°C
Color Colorless
pH Acidic
Physical Form Solid
Quantity 25 g
Linear Formula KHSO4
UN Number UN2509
Formula Weight 136.17g/mol
Percent Purity ≥99.995% (metals basis)
Grade TraceSELECT
Odor Odorless
Packaging Poly bottle
Density 2.32g/mL at 25°C
Chemical Name or Material Potassium bisulfate
Show More Show Less
Hazard Statement P261-P280-P305 + P351 + P338-P310
DOTInformation 8/PGII
RTECSNumber 231-594-1
Product Title
Select an issue

By clicking Submit, you acknowledge that you may be contacted by Fisher Scientific in regards to the feedback you have provided in this form. We will not share your information for any other purposes. All contact information provided shall also be maintained in accordance with our Privacy Policy.