Promotional price valid on web orders only. Your contract pricing may differ. Interested in signing up for a dedicated account number?
Learn More

Fisher Science Education™ Potassium Bitartrate
SCI_ED

Catalog No. S25477A Shop All Fisherbrand Products
Change view
Click to view available options
Quantity:
100 g
500 g
Packaging:
Reagent Grade
2 product options available for selection
Product selection table with 2 available options. Use arrow keys to navigate and Enter or Space to select.
Catalog No. Quantity Packaging
S25477A 500 g Reagent Grade
S25477 100 g Reagent Grade
Use arrow keys to navigate between rows. Press Enter or Space to select a product option. 2 options available.
2 options
Catalog No. S25477A

missing translation for 'validMainframeMsg'

This product is for educational use - no C of A is available.

Chemical Identifiers

CAS 868-14-4
Molecular Formula C4H5KO6
Molecular Weight (g/mol) 188.18
MDL Number MFCD00065392,MFCD00071626,MFCD00064206
InChI Key KYKNRZGSIGMXFH-ZVGUSBNCSA-M
Synonym potassium bitartrate, l +-potassium hydrogen tartrate, potassium 3-carboxy-2,3-dihydroxypropanoate, monopotassium tartrate, potassium acid tartrate, kaliumtartrat, potassium hydrogentartrate, 1-potassium hydrogentartrate, ksc657e1t, butanedioic acid, 2,3-dihydroxy-2r,3r-, potassium salt 1:1
PubChem CID 24193652
IUPAC Name potassium (2R,3R)-3-carboxy-2,3-dihydroxypropanoate
SMILES [K+].O[C@H]([C@@H](O)C([O-])=O)C(O)=O

Specifications

Color White
Density 1.984g/mL
Assay Percent Range 99% w/v
Packaging Reagent Grade
Linear Formula KHC4H4O6
Quantity 500 g
Formula Weight 188.18
Chemical Name or Material Potassium Bitartrate
Safety handling Check Alert storage code Red
ShelfLife 5 years
Product Title
Select an issue

By clicking Submit, you acknowledge that you may be contacted by Fisher Scientific in regards to the feedback you have provided in this form. We will not share your information for any other purposes. All contact information provided shall also be maintained in accordance with our Privacy Policy.