Promotional price valid on web orders only. Your contract pricing may differ. Interested in signing up for a dedicated account number?
Learn More

Potassium Bitartrate, USP, 99-101%, Spectrum™ Chemical
SDP

Catalog No. 18605468 Shop All Spectrum Chemical Mfg. Corp. Products
Change view
Click to view available options
Quantity:
500 g
2.5 kg
12 kg
50 kg
Packaging:
Poly Bottle
4 product options available for selection
Product selection table with 4 available options. Use arrow keys to navigate and Enter or Space to select.
Catalog No. Quantity Packaging
18-605-468 50 kg Poly Bottle
18-605-465 500 g Poly Bottle
18-605-466 2.5 kg Poly Bottle
18-605-467 12 kg Poly Bottle
Use arrow keys to navigate between rows. Press Enter or Space to select a product option. 4 options available.
4 options
Catalog No. 18-605-468 Supplier Spectrum Chemical Manufacturing Corporation Supplier No. P180150KGBL
Get Up to $300 in Additional Chemicals
View all promotions
Add to Cart
Add to Cart

missing translation for 'validMainframeMsg'

P1801, 868-14-4, C4H5KO6

Spectrum™ Chemical Potassium Bitartrate, USP is used internally as a purgative. All Spectrum Chemical USP grade products are manufactured, packaged and stored under current Good Manufacturing Practices (cGMP) per 21CFR part 211 in FDA registered and inspected facilities.

Chemical Identifiers

CAS 868-14-4
Molecular Formula C4H5KO6
Molecular Weight (g/mol) 188.18
MDL Number MFCD00065392,MFCD00071626,MFCD00064206
InChI Key KYKNRZGSIGMXFH-ZVGUSBNCSA-M
IUPAC Name potassium (2R,3R)-3-carboxy-2,3-dihydroxypropanoate
SMILES [K+].O[C@H]([C@@H](O)C([O-])=O)C(O)=O

Specifications

CAS Max % 1
Physical Form Crystalline Powder or Needles
Quantity 50 kg
Percent Purity 99 to 101%
Grade USP
Packaging Poly Bottle
Chemical Name or Material Potassium Bitartrate
Product Title
Select an issue

By clicking Submit, you acknowledge that you may be contacted by Fisher Scientific in regards to the feedback you have provided in this form. We will not share your information for any other purposes. All contact information provided shall also be maintained in accordance with our Privacy Policy.