Learn More
Potassium borohydride, 98%, Thermo Scientific™
Description
This Thermo Scientific brand product was originally part of the Alfa Aesar product portfolio. Some documentation and label information may refer to the legacy brand. The original Alfa Aesar product / item code or SKU reference has not changed as a part of the brand transition to Thermo Scientific.
- Source of hydrogen, reducing agent for aldehydes, ketones, acid chlorides; foaming agent for plastics
Specifications
Specifications
| Melting Point | >400°C (decomposition) |
| Physical Form | Granules |
| Quantity | 500 g |
| Linear Formula | KBH4 |
| UN Number | UN1870 |
| Merck Index | 14,7616 |
| Solubility Information | Soluble in water (19g/100g ); methanol (3;9g/100g ); 4:1 water:methanol mixture (13g/100g ). |
| IUPAC Name | potassium boranuide |
| Formula Weight | 53.94 |
| Percent Purity | 98% |
| Show More |
Safety and Handling
| Hazard Category | H261-H301+H311-H314 |
| Hazard Statement | GHS H Statement H261-H301-H311-H331-H314-H318 In contact with water releases flammable gas. Toxic if swallowed. Toxic in contact with skin. Toxic if inhaled. Causes severe skin burns and eye damage. Causes serious eye damage. |
| Precautionary Statement | P223-P231+P232-P260-P264b-P270-P280-P303+P361+P353-P304+P340-P305+P351+P338-P310-P330-P331-P335+P334-P363-P370+P378q-P501c |
| DOTInformation | Transport Hazard Class: 4.3; Packing Group: I; Proper Shipping Name: POTASSIUM BOROHYDRIDE |
| EINECSNumber | 237-360-5 |
| RTECSNumber | TS7525000 |
| TSCA | Yes |
| Recommended Storage | Ambient temperatures; Store under Nitrogen |
By clicking Submit, you acknowledge that you may be contacted by Fisher Scientific in regards to the feedback you have provided in this form. We will not share your information for any other purposes. All contact information provided shall also be maintained in accordance with our Privacy Policy.