Promotional price valid on web orders only. Your contract pricing may differ. Interested in signing up for a dedicated account number?
Learn More

Potassium Bromate, ACS, ≥99.8% (Dried material, RT), Solstice

Catalog No. 60047153 Shop All Solstice Research Chemicals Products
Change view
Click to view available options
Quantity:
50 g
250 g
Packaging:
Poly bottle
2 product options available for selection
Product selection table with 2 available options. Use arrow keys to navigate and Enter or Space to select.
Catalog No. Quantity Packaging
60-047-153 250 g Poly bottle
60-047-154 50 g Poly bottle
Use arrow keys to navigate between rows. Press Enter or Space to select a product option. 2 options available.
2 options
Catalog No. 60-047-153 Supplier Solstice Supplier No. 60085250G
Add to Cart
Add to Cart

missing translation for 'validMainframeMsg'

Puriss. p.a., ACS Reagent,≥99.8% (dried material, RT)

  • Leading the Industry For Over 65 Years
  • Solstice now delivers premium grade inorganic reagents worldwide – with consistency, purity, and accuracy assured
TRUSTED_SUSTAINABILITY

Chemical Identifiers

CAS 1-2-7758
Molecular Formula BrKO3
Molecular Weight (g/mol) 167.00
MDL Number MFCD00011359
InChI Key OCATYIAKPYKMPG-UHFFFAOYSA-M
Synonym potassium bromate, bromic acid, potassium salt, bromate, potassium, potassium bromate dot, unii-04mb35w6za, ccris 529, eec no. e924, bromic acid, potassium salt 1:1, potassiumbromate, kaliumbromat
PubChem CID 23673461
ChEBI CHEBI:38211
IUPAC Name potassium;bromate
SMILES [O-]Br(=O)=O.[K+]

Specifications

Color Colorless or White
pH 5.0 to 9.0 (at 25°C, 50mg/mL in H2O)
Physical Form Solid
Quantity 250 g
Linear Formula KBrO3
UN Number UN1484
Formula Weight 167g/mol
Percent Purity ≥99.8% (dried material, RT)
Grade ACS
Packaging Poly bottle
Density 3.270g/mL
Chemical Name or Material Potassium bromate
Show More Show Less
Hazard Statement P201-P220-P301 + P310-P308 + P313
DOTInformation 5.1 / PGII
RTECSNumber 231-829-8
WARNING: Cancer - www.P65Warnings.ca.gov
Product Title
Select an issue

By clicking Submit, you acknowledge that you may be contacted by Fisher Scientific in regards to the feedback you have provided in this form. We will not share your information for any other purposes. All contact information provided shall also be maintained in accordance with our Privacy Policy.