Promotional price valid on web orders only. Your contract pricing may differ. Interested in signing up for a dedicated account number?
Learn More

Potassium Bromate, FCC, 99-101%, Spectrum™ Chemical
SDP

Catalog No. 18605472 Shop All Spectrum Chemical Mfg. Corp. Products
Change view
Click to view available options
Quantity:
25 g
125 g
500 g
2.5 kg
Packaging:
Poly Bottle
Poly Pail
4 product options available for selection
Product selection table with 4 available options. Use arrow keys to navigate and Enter or Space to select.
Catalog No. Quantity Packaging
18-605-472 25 g Poly Bottle
18-605-469 125 g Poly Bottle
18-605-470 500 g Poly Bottle
18-605-471 2.5 kg Poly Pail
Use arrow keys to navigate between rows. Press Enter or Space to select a product option. 4 options available.
4 options
Catalog No. 18-605-472 Supplier Spectrum Chemical Manufacturing Corporation Supplier No. P127825GM
Get Up to $500 in Chemicals with Purchase
View all promotions
Add to Cart
Add to Cart

missing translation for 'validMainframeMsg'

P1278, 7758-01-2, KBrO3

Spectrum™ Chemical Potassium Bromate, FCC is considered an oxidizing agent and has been used in the production of malt barley. The FCC grade meets the requirements of the Food Chemical Codex indicates and is suitable for all food, beverage and nutritional supplement applications. Spectrum Chemical offers over 300 Food grade chemical ingredients packaged in laboratory size bottles to production drum quantities and are manufactured, packaged and stored under current Good Manufacturing Practices (cGMP) per 21CFR part 211 in FDA registered and inspected facilities.

Chemical Identifiers

CAS 1-2-7758
Molecular Formula BrKO3
Molecular Weight (g/mol) 167.00
InChI Key OCATYIAKPYKMPG-UHFFFAOYSA-M
IUPAC Name potassium bromate
SMILES [K+].[O-][Br](=O)=O

Specifications

CAS Max % 1
Physical Form Crystalline Powder
Quantity 25 g
Percent Purity 99 to 101%
Grade FCC
Loss on Drying 0.001
Packaging Poly Bottle
Chemical Name or Material Potassium Bromate
WARNING: Cancer - www.P65Warnings.ca.gov
Product Title
Select an issue

By clicking Submit, you acknowledge that you may be contacted by Fisher Scientific in regards to the feedback you have provided in this form. We will not share your information for any other purposes. All contact information provided shall also be maintained in accordance with our Privacy Policy.