Promotional price valid on web orders only. Your contract pricing may differ. Interested in signing up for a dedicated account number?
Learn More

Potassium bromide, 99+%, for spectroscopy, IR grade

Catalog No. p-222047
Change view
Click to view available options
Quantity:
25 g
100 g
500 g
Packaging:
Plastic bottle
3 product options available for selection
Product selection table with 3 available options. Use arrow keys to navigate and Enter or Space to select.
Catalog No. Quantity Packaging
AC206390250 25 g Plastic bottle
AC206391000 100 g Plastic bottle
AC206395000 500 g Plastic bottle
Use arrow keys to navigate between rows. Press Enter or Space to select a product option. 3 options available.
3 options
Catalog No. AC206390250 Supplier Thermo Scientific Chemicals Supplier No. 206390250
Only null left
Add to Cart

CAS: 7758-02-3 | BrK | 119.00 g/mol

This Thermo Scientific Chemicals brand product was originally part of the Acros Organics product portfolio. Some documentation and label information may refer to the legacy brand. The original Acros Organics product / item code or SKU reference has not changed as a part of the brand transition to Thermo Scientific Chemicals.

For IR spectroscopy
TRUSTED_SUSTAINABILITY

Chemical Identifiers

CAS 2-3-7758
Molecular Formula BrK
Molecular Weight (g/mol) 119.00
MDL Number MFCD00011358
InChI Key IOLCXVTUBQKXJR-UHFFFAOYSA-M
Synonym potassium bromide, bromide salt of potassium, kalii bromidum, bromure de potassium, caswell no. 684, tripotassium tribromide, potassium bromide k3br3, unii-osd78555zm, ccris 6095
PubChem CID 253877
ChEBI CHEBI:32030
SMILES [K+].[Br-]

Specifications

Boiling Point 1435.0°C
Melting Point 734.0°C
Color White
Physical Form Fine Crystalline Powder
Quantity 25 g
Assay Percent Range 99% min. (Argentometry)
Linear Formula KBr
Merck Index 15, 7739
Solubility Information Solubility in water: 650g/L (20°C). Other solubilities: 1020g/L water (100°C),1.42g/L ethanol (25°C),217g/L glycerol,4g/L alcohol
IUPAC Name potassium;bromide
Formula Weight 119
Percent Purity ≥99%
Grade Spectroscopy
Infrared Spectrum Authentic
Packaging Plastic bottle
Chemical Name or Material Potassium bromide, 99+%
Show More Show Less
Hazard Category GHS Signal Word: Warning
Hazard Statement GHS H Statement
Causes serious eye irritation.
Precautionary Statement GHS P Statement
Wear protective gloves/protective clothing/eye protection/face protection.
IF IN EYES: Rinse cautiously with water for several minutes.
Remove contact lenses,if present and easy to do.
Continue rinsing.
EINECSNumber 231-830-3
RTECSNumber TS7650000
TSCA TSCA

RUO – Research Use Only

Product Title
Select an issue

By clicking Submit, you acknowledge that you may be contacted by Fisher Scientific in regards to the feedback you have provided in this form. We will not share your information for any other purposes. All contact information provided shall also be maintained in accordance with our Privacy Policy.