Promotional price valid on web orders only. Your contract pricing may differ. Interested in signing up for a dedicated account number?
Learn More

Potassium bromide crystal optic disc, 25mm x 4mm, polished both sides

Catalog No. aa38425kt
Change view
Click to view available options
Quantity:
1 Ea.
1 product options available for selection
Product selection table with 1 available options. Use arrow keys to navigate and Enter or Space to select.
Catalog No. Quantity
AA38425KT 1 Ea.
Use arrow keys to navigate between rows. Press Enter or Space to select a product option. 1 options available.
1 options
Catalog No. AA38425KT Supplier Thermo Scientific Chemicals Supplier No. 038425KT
Only null left
Add to Cart

 

Potassium bromide crystal optic disc is used for infrared transmission windows in gas and liquid sample cells used with infrared and FTIR spectrophotometers and for beam splitters for spectrophotometers.

This Thermo Scientific Chemicals brand product was originally part of the Alfa Aesar product portfolio. Some documentation and label information may refer to the legacy brand. The original Alfa Aesar product / item code or SKU reference has not changed as a part of the brand transition to Thermo Scientific Chemicals.

TRUSTED_SUSTAINABILITY

Specifications

Chemical Name or Material Potassium bromide crystal optic disc
Name Note Polished on both sides
Quantity 1 Ea.
MDL Number MFCD00011358
Solubility Information Soluble in water,glycerol. Slightly soluble in ethanol and diethylether.

RUO – Research Use Only

Product Title
Select an issue

By clicking Submit, you acknowledge that you may be contacted by Fisher Scientific in regards to the feedback you have provided in this form. We will not share your information for any other purposes. All contact information provided shall also be maintained in accordance with our Privacy Policy.