Promotional price valid on web orders only. Your contract pricing may differ. Interested in signing up for a dedicated account number?
Learn More

Potassium bromide, FT-IR Grade, ≥99% trace metals basis, Solstice

Catalog No. 60045328 Shop All Solstice Research Chemicals Products
Change view
Click to view available options
Quantity:
25 g
100 g
2 product options available for selection
Product selection table with 2 available options. Use arrow keys to navigate and Enter or Space to select.
Catalog No. Quantity
60-045-328 25 g
60-021-31 100 g
Use arrow keys to navigate between rows. Press Enter or Space to select a product option. 2 options available.
2 options
Catalog No. 60-045-328 Supplier Solstice Supplier No. 22186425G
Add to Cart
Add to Cart

missing translation for 'validMainframeMsg'

Chemical Identifiers

CAS 2-3-7758
Molecular Formula BrK
Molecular Weight (g/mol) 119.00
MDL Number MFCD00011358
InChI Key IOLCXVTUBQKXJR-UHFFFAOYSA-M
Synonym potassium bromide, bromide salt of potassium, kalii bromidum, bromure de potassium, caswell no. 684, tripotassium tribromide, potassium bromide k3br3, unii-osd78555zm, ccris 6095
PubChem CID 253877
ChEBI CHEBI:32030
SMILES [K+].[Br-]

Specifications

Boiling Point 1,383°C
Melting Point 734°C (literature)
CAS Max % 100.0000%
Physical Form Powder
Quantity 25 g
Linear Formula KBr
UN Number NONH for all modes of transport
IUPAC Name potassium bromide
Formula Weight 119
Percent Purity >99%
Grade Not Available
Packaging Poly Bottle
Vapor Pressure 1mmHg (795°C), 0.01mmHg (20°C)
Density 2.73 g/cm3
Chemical Name or Material Potassium bromide, FT-IR Grade, ≥99% trace metals basis
Show More Show Less
Hazard Statement P280-P305 + P351 + P338-P337 + P313
Precautionary Statement H319
ShelfLife 1080 days from date of manufacture
EINECSNumber 231-830-3
RTECSNumber TS7650000
Recommended Storage Room Temp
WARNING: Reproductive Harm - www.P65Warnings.ca.gov
Product Title
Select an issue

By clicking Submit, you acknowledge that you may be contacted by Fisher Scientific in regards to the feedback you have provided in this form. We will not share your information for any other purposes. All contact information provided shall also be maintained in accordance with our Privacy Policy.