Promotional price valid on web orders only. Your contract pricing may differ. Interested in signing up for a dedicated account number?
Learn More

Potassium bromide, Premium FTIR Grade

Catalog No. p-7052261
Change view
Click to view available options
Quantity:
100 Pc.
200 Pc.
25 Pc.
50 Pc.
4 product options available for selection
Product selection table with 4 available options. Use arrow keys to navigate and Enter or Space to select.
Catalog No. Quantity
AA47111KM 25 Pc.
AA47111KD 50 Pc.
AA47111KN 100 Pc.
AA47111KQ 200 Pc.
Use arrow keys to navigate between rows. Press Enter or Space to select a product option. 4 options available.
4 options
Catalog No. AA47111KM Supplier Thermo Scientific Chemicals Supplier No. 047111KM
Only null left
Add to Cart

CAS: 7758-02-3 | BrK | 119.00 g/mol

This Thermo Scientific Chemicals brand product was originally part of the Alfa Aesar product portfolio. Some documentation and label information may refer to the legacy brand. The original Alfa Aesar product / item code or SKU reference has not changed as a part of the brand transition to Thermo Scientific Chemicals.

TRUSTED_SUSTAINABILITY

Chemical Identifiers

CAS 2-3-7758
Molecular Formula BrK
Molecular Weight (g/mol) 119.00
MDL Number MFCD00011358
InChI Key IOLCXVTUBQKXJR-UHFFFAOYSA-M
Synonym potassium bromide, bromide salt of potassium, kalii bromidum, bromure de potassium, caswell no. 684, tripotassium tribromide, potassium bromide k3br3, unii-osd78555zm, ccris 6095
PubChem CID 253877
ChEBI CHEBI:32030
IUPAC Name potassium bromide
SMILES [K+].[Br-]

Specifications

Boiling Point 1435°C
Melting Point 730°C
Physical Form Powder
Quantity 25 Pc.
Linear Formula KBr
Merck Index 14,7618
Formula Weight 119.01
Grade Premium FTIR
Odor Odorless
Packaging 0.5g packets
Refractive Index 1.559
Density 2.75 g/mL
Chemical Name or Material Potassium bromide
Show More Show Less
Hazard Category H319-H335
Hazard Statement GHS H Statement
H315-H319-H335
Causes skin irritation.
Causes serious eye irritation.
May cause respiratory irritation.
Precautionary Statement P261-P264b-P271-P280i-P304+P340-P305+P351+P338-P312-P501c
EINECSNumber 231-830-3
RTECSNumber TS7650000
TSCA Yes
Recommended Storage Ambient temperatures

RUO – Research Use Only

Product Title
Select an issue

By clicking Submit, you acknowledge that you may be contacted by Fisher Scientific in regards to the feedback you have provided in this form. We will not share your information for any other purposes. All contact information provided shall also be maintained in accordance with our Privacy Policy.