Promotional price valid on web orders only. Your contract pricing may differ. Interested in signing up for a dedicated account number?
Learn More

Potassium Bromide, puriss., Solstice

Catalog No. 60044466 Shop All Solstice Research Chemicals Products
Change view
Click to view available options
Quantity:
100 g
500 g
1 kg
2.5 kg
6 x 1 kg
5 product options available for selection
Product selection table with 5 available options. Use arrow keys to navigate and Enter or Space to select.
Catalog No. Quantity
60-044-466 6 x 1 kg
60-017-95 100 g
60-017-99 500 g
60-017-96 1 kg
60-017-97 2.5 kg
Use arrow keys to navigate between rows. Press Enter or Space to select a product option. 5 options available.
5 options
Catalog No. 60-044-466 Supplier Solstice Supplier No. 021106X1KG
Add to Cart
Add to Cart

missing translation for 'validMainframeMsg'

Puriss., meets analytical specification of BP, Ph. Eur., USP, 99.5-100.5%

  • Leading the Industry For Over 65 Years
  • Solstice now delivers premium grade inorganic reagents worldwide – with consistency, purity, and accuracy assured
TRUSTED_SUSTAINABILITY

Chemical Identifiers

Molecular Formula BrK
Molecular Weight (g/mol) 119.00
MDL Number MFCD00011358
InChI Key IOLCXVTUBQKXJR-UHFFFAOYSA-M
Synonym potassium bromide, bromide salt of potassium, kalii bromidum, bromure de potassium, caswell no. 684, tripotassium tribromide, potassium bromide k3br3, unii-osd78555zm, ccris 6095
PubChem CID 253877
ChEBI CHEBI:32030
SMILES [K+].[Br-]

Specifications

CAS 2-3-7758
Quantity 6 x 1 kg
Linear Formula KBr
UN Number NONH for all modes of transport
IUPAC Name potassium;bromide
Formula Weight 119
Percent Purity 99.5 to 100.5%
Grade puriss.
Vapor Pressure 0.01mmHg (20°C), 1mmHg (795°C)
Chemical Name or Material Potassium Bromide, Meets analytical specification of Ph. Eur.BP,USP
Hazard Statement P280-P305 + P351 + P338-P337 + P313
Precautionary Statement H319
EINECSNumber 231-830-3
RTECSNumber TS7650000
WARNING: Cancer - www.P65Warnings.ca.gov
Product Title
Select an issue

By clicking Submit, you acknowledge that you may be contacted by Fisher Scientific in regards to the feedback you have provided in this form. We will not share your information for any other purposes. All contact information provided shall also be maintained in accordance with our Privacy Policy.