Promotional price valid on web orders only. Your contract pricing may differ. Interested in signing up for a dedicated account number?
Learn More

Potassium Bromide, puriss p.a, ACS, ≥99.5% (AT), Solstice

Catalog No. 60047155 Shop All Solstice Research Chemicals Products
Change view
Click to view available options
Quantity:
250 g
1 kg
Packaging:
Poly bottle
2 product options available for selection
Product selection table with 2 available options. Use arrow keys to navigate and Enter or Space to select.
Catalog No. Quantity Packaging
60-047-155 1 kg Poly bottle
60-047-156 250 g Poly bottle
Use arrow keys to navigate between rows. Press Enter or Space to select a product option. 2 options available.
2 options
Catalog No. 60-047-155 Supplier Solstice Supplier No. 600931KG
Add to Cart
Add to Cart

missing translation for 'validMainframeMsg'

Puriss. p.a., ACS Reagent,≥99.5% (AT)

  • Leading the Industry For Over 65 Years
  • Solstice now delivers premium grade inorganic reagents worldwide – with consistency, purity, and accuracy assured
TRUSTED_SUSTAINABILITY

Chemical Identifiers

CAS 2-3-7758
Molecular Formula BrK
Molecular Weight (g/mol) 119.00
MDL Number MFCD00011358
InChI Key IOLCXVTUBQKXJR-UHFFFAOYSA-M
Synonym potassium bromide, bromide salt of potassium, kalii bromidum, bromure de potassium, caswell no. 684, tripotassium tribromide, potassium bromide k3br3, unii-osd78555zm, ccris 6095
PubChem CID 253877
ChEBI CHEBI:32030
IUPAC Name potassium;bromide
SMILES [K+].[Br-]

Specifications

Boiling Point 1383°C
Melting Point 734°C
Color Colorless or White
pH 5.0 to 8.8
Quantity 1 kg
Linear Formula KBr
UN Number NONH for all modes of transport
Formula Weight 119g/mol
Percent Purity ≥99.5% (AT)
Grade ACS
Packaging Poly bottle
Vapor Pressure 1mmHg (795°C), 0.01mmHg (20°C)
Density 2.73g/mL
Chemical Name or Material Potassium bromide
Show More Show Less
Hazard Statement P280-P305 + P351 + P338-P337 + P313
RTECSNumber 231-830-3
WARNING: Reproductive Harm - www.P65Warnings.ca.gov
Product Title
Select an issue

By clicking Submit, you acknowledge that you may be contacted by Fisher Scientific in regards to the feedback you have provided in this form. We will not share your information for any other purposes. All contact information provided shall also be maintained in accordance with our Privacy Policy.