Promotional price valid on web orders only. Your contract pricing may differ. Interested in signing up for a dedicated account number?
Learn More

Potassium Bromide, TraceSELECT™, ≥99.99% (Metals Basis), Solstice

Catalog No. 60047742 Shop All Honeywell Research Chemicals Products
Change view
Click to view available options
Quantity:
25 g
100 g
Packaging:
Poly Bottle
2 product options available for selection
Product selection table with 2 available options. Use arrow keys to navigate and Enter or Space to select.
Catalog No. Quantity Packaging
60-047-742 25 g Poly Bottle
60-047-741 100 g Poly Bottle
Use arrow keys to navigate between rows. Press Enter or Space to select a product option. 2 options available.
2 options
Catalog No. 60-047-742 Supplier Solstice Supplier No. 9073725G

Chemical Identifiers

CAS 2-3-7758
Molecular Formula BrK
Molecular Weight (g/mol) 119.00
MDL Number MFCD00011358
InChI Key IOLCXVTUBQKXJR-UHFFFAOYSA-M
Synonym potassium bromide, bromide salt of potassium, kalii bromidum, bromure de potassium, caswell no. 684, tripotassium tribromide, potassium bromide k3br3, unii-osd78555zm, ccris 6095
PubChem CID 253877
ChEBI CHEBI:32030
IUPAC Name potassium bromide
SMILES [K+].[Br-]

Specifications

Boiling Point 1383°C
Melting Point 734°C
Color Colorless
Quantity 25 g
Linear Formula KBr
UN Number NONH for all modes of transport
Solubility Information 656g/L at 20°C
Formula Weight 119g/mol
Percent Purity ≥99.9995% (metals basis)
Grade TraceSELECT
Odor Odorless
Packaging Poly Bottle
Appearance Solid
Vapor Pressure 0.01mmHg (20°C), 1mmHg (795°C)
Density In water 2.73g/cm3 at 20°C
Chemical Name or Material Potassium bromide
Show More Show Less
Hazard Statement P280-P305 + P351 + P338-P337 + P313
RTECSNumber 231-830-3
Product Title
Select an issue

By clicking Submit, you acknowledge that you may be contacted by Fisher Scientific in regards to the feedback you have provided in this form. We will not share your information for any other purposes. All contact information provided shall also be maintained in accordance with our Privacy Policy.