Promotional price valid on web orders only. Your contract pricing may differ. Interested in signing up for a dedicated account number?
Learn More

Potassium bromide, ultra dry, 99.95% (metals basis)

Catalog No. AA4128222
Change view
Click to view available options
Quantity:
25 g
100 g
2 product options available for selection
Product selection table with 2 available options. Use arrow keys to navigate and Enter or Space to select.
Catalog No. Quantity
AA4128222 100 g
AA4128214 25 g
Use arrow keys to navigate between rows. Press Enter or Space to select a product option. 2 options available.
2 options
Catalog No. AA4128222 Supplier Thermo Scientific Chemicals Supplier No. 04128222
Add to Cart

missing translation for 'validMainframeMsg'

CAS: 7758-02-3 | BrK | 119.00 g/mol

Potassium bromide is used in a variety of applications such as spectroscopy, infrared transmission, as a laboratory reagent, and for the removal of peripheral membrane proteins. It is also used in photosensitive chemicals, paints and coatings.

This Thermo Scientific Chemicals brand product was originally part of the Alfa Aesar product portfolio. Some documentation and label information may refer to the legacy brand. The original Alfa Aesar product / item code or SKU reference has not changed as a part of the brand transition to Thermo Scientific Chemicals.

TRUSTED_SUSTAINABILITY

Chemical Identifiers

CAS 2-3-7758
Molecular Formula BrK
Molecular Weight (g/mol) 119.00
MDL Number MFCD00011358
InChI Key IOLCXVTUBQKXJR-UHFFFAOYSA-M
Synonym potassium bromide, bromide salt of potassium, kalii bromidum, bromure de potassium, caswell no. 684, tripotassium tribromide, potassium bromide k3br3, unii-osd78555zm, ccris 6095
PubChem CID 253877
ChEBI CHEBI:32030
SMILES [K+].[Br-]

Specifications

Boiling Point 1435°C
Melting Point 730°C
Color White
Physical Form Powder
Quantity 100 g
Assay Percent Range 99.95% (metals basis)
Solubility Information Soluble in water. Slightly soluble in alcohol,glycerol and ether.
IUPAC Name potassium;bromide
Formula Weight 119.01
Odor Odorless
Refractive Index 1.559
Sensitivity Hygroscopic
Appearance White Powder/crystals
Density 2.75 g/cm3 at 20°C
Chemical Name or Material Potassium bromide, Ampouled under argon
Show More Show Less
Hazard Category H319-H335
Precautionary Statement P261-P264b-P271-P280i-P304+P340-P305+P351+P338-P312-P501c
EINECSNumber 231-830-3
RTECSNumber TS7650000
TSCA Yes
Recommended Storage Ambient temperatures; Store under Nitrogen

RUO – Research Use Only

Product Title
Select an issue

By clicking Submit, you acknowledge that you may be contacted by Fisher Scientific in regards to the feedback you have provided in this form. We will not share your information for any other purposes. All contact information provided shall also be maintained in accordance with our Privacy Policy.