Learn More
Medchemexpress LLC Potassium (bromomethyl)trifluoroborate | 888711-44-2 | MFCD09265154 | 200.84 g/mol | BrCH2BF3K | 500 G
SDP
Catalog No. 5000302120
Encompass_Preferred
Potassium (bromomethyl)trifluoroborate (CAS 888711-44-2) is an organoboron reagent used as a boron-containing building block in organic synthesis. It is provided as a stable potassium trifluoroborate salt that simplifies handling and storage compared with some boronic acids, and is commonly employed in palladium-catalyzed transformations and modular assembly of complex molecules.
  • Organoboron building block for cross-coupling reactions.
  • Potassium trifluoroborate salt provides enhanced stability and ease of handling.
  • Solid form suitable for accurate weighing and storage.
  • Compatible with typical palladium-catalyzed conditions used in synthesis.
  • Supplied with a specified purity; verify certificate of analysis before use.
Catalog No. 50-003-02120 Supplier Medchemexpress LLC Supplier No. HYW014866500G
Potassium (bromomethyl)trifluoroborate (CAS 888711-44-2) is an organoboron reagent used as a boron-containing building block in organic synthesis. It is provided as a stable potassium trifluoroborate salt that simplifies handling and storage compared with some boronic acids, and is commonly employed in palladium-catalyzed transformations and modular assembly of complex molecules.
  • Organoboron building block for cross-coupling reactions.
  • Potassium trifluoroborate salt provides enhanced stability and ease of handling.
  • Solid form suitable for accurate weighing and storage.
  • Compatible with typical palladium-catalyzed conditions used in synthesis.
  • Supplied with a specified purity; verify certificate of analysis before use.