Learn More
Medchemexpress LLC Potassium (bromomethyl)trifluoroborate | 888711-44-2 | MFCD09265154 | 90.0% | 200.84 | CH2BBrF3K | 1 KG
SDP
Catalog No. 5000302366
Encompass_Preferred
Potassium (bromomethyl)trifluoroborate is an organotrifluoroborate salt used as a building block in organic synthesis. It is intended for laboratory research use and provides a convenient source of bromomethyltrifluoroborate functionality for cross-coupling and related transformations.
  • Used as a building block for Suzuki-Miyaura and other cross-coupling reactions.
  • Stable, easy-to-handle solid suitable for bench use.
  • Compatible with aqueous and polar reaction media.
  • Appropriate for laboratory-scale synthesis and method development.
  • Provided with an SDS to support safe handling and regulatory compliance.
Catalog No. 50-003-02366 Supplier Medchemexpress LLC Supplier No. HYW0148661KG
Potassium (bromomethyl)trifluoroborate is an organotrifluoroborate salt used as a building block in organic synthesis. It is intended for laboratory research use and provides a convenient source of bromomethyltrifluoroborate functionality for cross-coupling and related transformations.
  • Used as a building block for Suzuki-Miyaura and other cross-coupling reactions.
  • Stable, easy-to-handle solid suitable for bench use.
  • Compatible with aqueous and polar reaction media.
  • Appropriate for laboratory-scale synthesis and method development.
  • Provided with an SDS to support safe handling and regulatory compliance.