Learn More
Medchemexpress LLC Potassium (bromomethyl)trifluoroborate | 888711-44-2 | MFCD09265154 | 97.0% | 200.84 g·mol⁻¹ | BrCH2BF3K | 25 G
SDP
Catalog No. 5000303586
Encompass_Preferred
Potassium (bromomethyl)trifluoroborate is an organotrifluoroborate salt used as an organoboron reagent in organic synthesis and medicinal chemistry research. It functions as a stable, isolable boron-containing building block for cross-coupling and functionalization reactions and is supplied for research use only.
  • Stable organotrifluoroborate salt suitable for handling and storage.
  • Provides a bromomethyl boron synthon for cross-coupling reactions.
  • Compatible with common palladium-catalyzed coupling protocols.
  • Useful as a building block in medicinal and synthetic chemistry research.
  • Supplied in small laboratory quantities for research applications.
Catalog No. 50-003-03586 Supplier Medchemexpress LLC Supplier No. 25GHYW014866
Potassium (bromomethyl)trifluoroborate is an organotrifluoroborate salt used as an organoboron reagent in organic synthesis and medicinal chemistry research. It functions as a stable, isolable boron-containing building block for cross-coupling and functionalization reactions and is supplied for research use only.
  • Stable organotrifluoroborate salt suitable for handling and storage.
  • Provides a bromomethyl boron synthon for cross-coupling reactions.
  • Compatible with common palladium-catalyzed coupling protocols.
  • Useful as a building block in medicinal and synthetic chemistry research.
  • Supplied in small laboratory quantities for research applications.