Learn More
Medchemexpress LLC Potassium (bromomethyl)trifluoroborate | 888711-44-2 | MFCD09265154 | ≥97.0% | 200.84 g/mol | CH2BBrF3K | 1 G
SDP
Catalog No. 5000302599
Encompass_Preferred
Potassium (bromomethyl)trifluoroborate is a potassium organotrifluoroborate reagent used as a building block in organic synthesis and life-science research. It is typically supplied as a white to off-white powder for laboratory use and is applied in cross-coupling reactions and related transformations.
  • High purity suitable for research applications (NMR ≥97.0%).
  • Molecular formula CH2BBrF3K; molecular weight ~200.84 g/mol.
  • White to off-white powder for convenient handling and storage.
  • Commonly used as a coupling partner in cross-coupling and synthetic transformations.
  • Supplied with batch COA and SDS for quality and safety information.
Catalog No. 50-003-02599 Supplier Medchemexpress LLC Supplier No. HYW0148661G
Potassium (bromomethyl)trifluoroborate is a potassium organotrifluoroborate reagent used as a building block in organic synthesis and life-science research. It is typically supplied as a white to off-white powder for laboratory use and is applied in cross-coupling reactions and related transformations.
  • High purity suitable for research applications (NMR ≥97.0%).
  • Molecular formula CH2BBrF3K; molecular weight ~200.84 g/mol.
  • White to off-white powder for convenient handling and storage.
  • Commonly used as a coupling partner in cross-coupling and synthetic transformations.
  • Supplied with batch COA and SDS for quality and safety information.