Promotional price valid on web orders only. Your contract pricing may differ. Interested in signing up for a dedicated account number?
Learn More

Medchemexpress LLC Potassium (bromomethyl)trifluoroborate | 888711-44-2 | MFCD09265154 | ≥97.0% | 200.84 g/mol | CH2BBrF3K | 5 G
SDP

Catalog No. 5000303273
Encompass_Preferred

Potassium (bromomethyl)trifluoroborate is an organotrifluoroborate reagent used in organic synthesis and life-science research. It is commonly employed in cross-coupling and labeling reactions where a stable boron-containing nucleophile is required. Supplied as a solid for laboratory research, handle under anhydrous conditions and store sealed to minimize moisture exposure.

  • High analytical purity (≥97.0% by NMR)
  • Molecular formula CH2BBrF3K
  • Molecular weight 200.84 g/mol
  • Solid powder suitable for small-scale synthesis
  • Store sealed, away from moisture; frozen solutions for long-term storage

Catalog No. 50-003-03273 Supplier Medchemexpress LLC Supplier No. HYW0148665G
May include imposed supplier surcharges.
Add to Cart
Add to Cart

missing translation for 'validMainframeMsg'

Potassium (bromomethyl)trifluoroborate is an organotrifluoroborate reagent used in organic synthesis and life-science research. It is commonly employed in cross-coupling and labeling reactions where a stable boron-containing nucleophile is required. Supplied as a solid for laboratory research, handle under anhydrous conditions and store sealed to minimize moisture exposure.

  • High analytical purity (≥97.0% by NMR)
  • Molecular formula CH2BBrF3K
  • Molecular weight 200.84 g/mol
  • Solid powder suitable for small-scale synthesis
  • Store sealed, away from moisture; frozen solutions for long-term storage

Product Title
Select an issue

By clicking Submit, you acknowledge that you may be contacted by Fisher Scientific in regards to the feedback you have provided in this form. We will not share your information for any other purposes. All contact information provided shall also be maintained in accordance with our Privacy Policy.