Promotional price valid on web orders only. Your contract pricing may differ. Interested in signing up for a dedicated account number?
Learn More

Potassium Carbonate-1.5-hydrate, Suprapur™, MilliporeSigma™

Catalog No. M1049260050 Shop All MilliporeSigma Products
Change view
Click to view available options
Quantity:
50 g
Packaging:
Plastic Bottle
1 product options available for selection
Product selection table with 1 available options. Use arrow keys to navigate and Enter or Space to select.
Catalog No. Quantity Packaging
M1049260050 50 g Plastic Bottle
Use arrow keys to navigate between rows. Press Enter or Space to select a product option. 1 options available.
1 options
Catalog No. M1049260050 Supplier MilliporeSigma Supplier No. 1.04926.0050
Add to Cart
Add to Cart

Chemical Identifiers

CAS 6381-79-9
Molecular Formula C2H6K4O9
Molecular Weight (g/mol) 330.45
MDL Number MFCD00149925
InChI Key WWVGGBSGYNQKCY-UHFFFAOYSA-J
IUPAC Name tetrapotassium trihydrate dicarbonate
SMILES O.O.O.[K+].[K+].[K+].[K+].[O-]C([O-])=O.[O-]C([O-])=O

Specifications

Melting Point 891°C (anhydrous substance)
Color White
pH Strongly Alkaline
Physical Form Solid
Quantity 50 g
Solubility Information 1300g/L
Formula Weight 138.2
Percent Purity >99.995%
Packaging Plastic Bottle
EINECSNumber 209-529-3
Product Title
Select an issue

By clicking Submit, you acknowledge that you may be contacted by Fisher Scientific in regards to the feedback you have provided in this form. We will not share your information for any other purposes. All contact information provided shall also be maintained in accordance with our Privacy Policy.