Promotional price valid on web orders only. Your contract pricing may differ. Interested in signing up for a dedicated account number?
Learn More

Potassium Carbonate, 15% w/v, Reagents

Catalog No. 01334201
Change view
Click to view available options
Quantity: 500 mL
1 product options available for selection
Product selection table with 1 available options. Use arrow keys to navigate and Enter or Space to select.
Catalog No.
01-334-201 Quantity: 500 mL
Use arrow keys to navigate between rows. Press Enter or Space to select a product option. 1 options available.
1 options
Catalog No. 01-334-201 Supplier Reagents Holdings Llc Supplier No. CP202800500A
 Restricted Item
Add to Cart
Add to Cart

Potassium Carbonate, 15% w/v, Reagent, for general laboratory use. Manufactured in ISO 9001 facility.

Chemical Identifiers

CAS 7732-18-5 , 584-08-7
Molecular Formula H2O
Molecular Weight (g/mol) 18.02
InChI Key XLYOFNOQVPJJNP-UHFFFAOYSA-N
Synonym Dipotassium Carbonate, Carbonate of Potash
IUPAC Name water
SMILES O

Specifications

Boiling Point 100°C
Color Colorless
pH 11.6
Physical Form Liquid
Quantity 500 mL
Percent Purity 15%
Grade Reagent
Packaging Poly Bottle
Specific Gravity 1.13
Chemical Name or Material Potassium Carbonate, 15% w/v
Product Title
Select an issue

By clicking Submit, you acknowledge that you may be contacted by Fisher Scientific in regards to the feedback you have provided in this form. We will not share your information for any other purposes. All contact information provided shall also be maintained in accordance with our Privacy Policy.