Promotional price valid on web orders only. Your contract pricing may differ. Interested in signing up for a dedicated account number?
Learn More

Potassium Carbonate, 15% (w/v), Ricca Chemical
SDP

Catalog No. p-7239396 Shop All Ricca Chemical Company Products
Change view
Click to view available options
Quantity:
120 mL
500 mL
Packaging:
Natural Poly Bottle
2 product options available for selection
Product selection table with 2 available options. Use arrow keys to navigate and Enter or Space to select.
Catalog No. Quantity Packaging
R5883150120 120 mL Natural Poly Bottle
588315-16 500 mL Natural Poly Bottle
Use arrow keys to navigate between rows. Press Enter or Space to select a product option. 2 options available.
2 options
Catalog No. R5883150120 Supplier Ricca Chemical Company Supplier No. R5883150120A
Add to Cart
Add to Cart

Chemical Identifiers

CAS 584-08-7 , 7732-18-5
Molecular Formula CK2O3
Molecular Weight (g/mol) 138.21
MDL Number MFCD00011382
InChI Key BWHMMNNQKKPAPP-UHFFFAOYSA-L
IUPAC Name dipotassium carbonate
SMILES [K+].[K+].[O-]C([O-])=O

Specifications

Color Colorless Liquid
CAS Max % 13.27, 86.73
Concentration 14.6 to 15.4 N
Percent Purity 15%
Grade Laboratory
Packaging Natural Poly Bottle
pH >12
Quantity 120 mL
Sterility Non-sterile
Chemical Name or Material Potassium Carbonate
ShelfLife 12 months
Product Title
Select an issue

By clicking Submit, you acknowledge that you may be contacted by Fisher Scientific in regards to the feedback you have provided in this form. We will not share your information for any other purposes. All contact information provided shall also be maintained in accordance with our Privacy Policy.